Learn More
TNFRSF8 Mouse anti-Human, Clone: Ki-1/779, Abnova™
Mouse monoclonal antibody raised against full length recombinant human TNFRSF8.
Supplier: Abnova Corporation MAB13464
Description
The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is expressed by activated, but not by resting, T and B cells. TRAF2 and TRAF5 can interact with this receptor, and mediate the signal transduction that leads to the activation of NF-kappaB. This receptor is a positive regulator of apoptosis, and also has been shown to limit the proliferative potential of autoreactive CD8 effector T cells and protect the body against autoimmunity. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq]
Specifications
TNFRSF8 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
CD30/D1S166E/KI-1 | |
Mouse | |
Protein A/G purification | |
RUO | |
943 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
Ki-1/779 | |
Mouse monoclonal antibody raised against full length recombinant human TNFRSF8. | |
In 10mM PBS. | |
TNFRSF8 | |
Recombinant protein corresponding to full length human TNFRSF8. | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.