Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF9 Mouse anti-Human, Clone:4G10B9, Abnova™

Catalog No. 89334505 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-505 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-505 Supplier Abnova Corporation Supplier No. MAB17862
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human TNFRSF9.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor contributes to the clonal expansion, survival, and development of T cells. It can also induce proliferation in peripheral monocytes, enhance T cell apoptosis induced by TCR/CD3 triggered activation, and regulate CD28 co-stimulation to promote Th1 cell responses. The expression of this receptor is induced by lymphocyte activation. TRAF adaptor proteins have been shown to bind to this receptor and transduce the signals leading to activation of NF-kappaB. [provided by RefSeq]

Specifications

Antigen TNFRSF9
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4G10B9
Conjugate Unconjugated
Description tumor necrosis factor receptor superfamily, member 9
Dilution ELISA (1:10000)
Flow Cytometry (1:200-400)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene TNFRSF9
Gene Alias 4-1BB, CD137, CDw137, ILA, MGC2172
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 24-186 of human TNFRSF9 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 3604
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.