Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFRSF9 Mouse anti-Human, Clone: OTI7F9, lyophilized, TrueMAB™
SDP

Catalog No. 501665722
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-5722 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-5722 Supplier OriGene Supplier No. CF801767
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

CD137 is a member of the TNF-receptor superfamily and contributes to the clonal expansion, survival, and development of T cells. CD137 exists as both a monomer and a dimer on the surface of activated T cells. CD137 can also induce proliferation in peripheral monocytes, enhance T cell apoptosis induced by TCR/CD3 triggered activation, and regulate CD28 co-stimulation to promote Th1 cell responses. The expression of CD137 is induced by lymphocyte activation. CD137 is also an inducible receptor-like protein expressed on the cell surface of activated splenic T cells and thymocytes. CD137 exists as both a monomer and a dimer on the surface of activated T cells.

Specifications

Antigen TNFRSF9
Applications Western Blot
Classification Monoclonal
Clone OTI7F9
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene TNFRSF9
Gene Accession No. Q07011
Gene Alias 4-1BB, CD137, CDw137, ILA
Gene Symbols TNFRSF9
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 24-186 of human TNFRSF9 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3604
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.