Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFSF13B Mouse anti-Human, Clone: C257/1638, Abnova™

Catalog No. 89034809 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-809 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-809 Supplier Abnova Corporation Supplier No. MAB14726
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant human TNFSF13B.

  • High specificity
  • High reactivity

Specifications

Antigen TNFSF13B
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone C257/1638
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human TNFSF13B.
Dilution Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS.
Gene Accession No. Q9Y275
Gene Alias BAFF/BLYS/CD257/DTL/TALL-1/TALL1/THANK/TNFSF20/ZTNF4
Gene Symbols TNFSF13B
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human TNFSF13B.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10673
Target Species Human
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.