Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFSF13B, Rat anti-Human, Clone: 1-35-1, Abnova™

Catalog No. 89998879 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-879 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-879 Supplier Abnova Corporation Supplier No. MAB16104
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifications

Antigen TNFSF13B
Applications Flow Cytometry
Classification Monoclonal
Clone 1-35-1
Conjugate Unconjugated
Dilution Flow Cytometry The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide).
Gene tumor necrosis factor (ligand) superfamily, member 13b
Gene Alias BAFF/BLYS/CD257/DTL/TALL-1/TALL1/THANK/TNFSF20/ZTNF4
Gene Symbols TNFSF13B
Host Species Rat
Immunogen HA-tagged human TNFSF13B.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10673
Target Species Human
Content And Storage For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.