Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFSF15 Rabbit anti-Human, Unconjugated, Clone: VEGI/2052R, Abnova™

Catalog No. 89339723 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-723 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-723 Supplier Abnova Corporation Supplier No. RAB00684
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

TNFSF15 Monoclonal antibody specifically detects TNFSF15 in Human samples. It is validated for IHC-P.

Specifications

Antigen TNFSF15
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone VEGI/2052R
Conjugate Unconjugated
Description tumor necrosis factor (ligand) superfamily, member 15
Gene TNFSF15
Gene Alias MGC129934, MGC129935, TL1, TL1A, VEGI, VEGI192A
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human TNFSF15
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 9966
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.