Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFSF15 Rat anti-Mouse, Clone: Tan 2-2, MilliporeSigma™

Catalog No. MABC1628100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1628100 100 μg
MABC162825 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1628100 Supplier MilliporeSigma Supplier No. MABC1628100UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TNFSF15 Monoclonal antibody specifically detects TNFSF15 in Mouse samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry, Neutralizing, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFSF15
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Neutralization, Western Blot
Classification Monoclonal
Clone Tan 2-2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal antibody IgG2a in PBS without preservatives.
Host Species Rat
Immunogen Recombinant fragment corresponding to 176 amino acids from the extracellular domain of murine TNFSF15 TL1A protein with a human IgG1 Fc domain and an additional hinge-like region at the C-terminus.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.