Learn More
Description
Specifications
Specifications
| Antigen | TNFSF15 |
| Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry, Neutralization, Western Blot |
| Classification | Monoclonal |
| Clone | Tan 2-2 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Purified rat monoclonal antibody IgG2a in PBS without preservatives. |
| Host Species | Rat |
| Immunogen | Recombinant fragment corresponding to 176 amino acids from the extracellular domain of murine TNFSF15 TL1A protein with a human IgG1 Fc domain and an additional hinge-like region at the C-terminus. |
| Purification Method | Protein G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.