Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFSF4, anti-Human, Clone: R4930 (Oxelumab), Abnova™

Catalog No. 89032393 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-393 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-393 Supplier Abnova Corporation Supplier No. RAB00050
Add to Cart
Add to Cart

Monoclonal Antibody

Specifications

Antigen TNFSF4
Applications Blocking Assay, Flow Cytometry
Classification Monoclonal
Clone R4930 (Oxelumab)
Conjugate Unconjugated
Dilution Blocking Flow Cytometry (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.02% Proclin 300).
Gene tumor necrosis factor (ligand) superfamily, member 4
Gene Alias CD134L/CD252/GP34/OX-40L/OX4OL/TXGP1
Gene Symbols TNFSF4
Immunogen Original antibody is raised against human TNFSF4.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7292
Test Specificity This antibody binds to human TNFSF4.
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.