Learn More
Promega TNIK Kinase Enzyme System, 10microg
Catalog No. PRV4158 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4158 10 μg
1 options
Catalog No. PRV4158 Supplier Promega Supplier No. V4158
Recombinant human TNIK (1-367) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. TNIK or TRAF2 and NCK-interacting kinase are characterized by N-terminal kinase and C-terminal GCK domains that serve a regulatory function.
  • Easily Screen and Profile TNIK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C