Learn More
Invitrogen™ TNIK Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533085
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Bat, Rabbit (100%); Dog, Elephant, Panda, Horse (94%); Mouse, Rat (89%); Bovine (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
TNIK or TRAF2 and NCK interacting kinase is characterized by an N-terminal kinase domain and a C-terminal GCK domain that serves a regulatory function. TNIK is mainly expression in brain, heart, and spleen and it is a specific effector of RAP2 which regulate actin cytoskeleton. TNIK is autophosphorylated in a manner dependent upon lys54 in the ATP-binding pocket of its kinase domain and plays a main role in cytoskeleton regulation. TNIK has also been shown to phosphorylate gelsolin, the principal intracellular and extracellular actin-severing protein, in vitro. This and evidence from mutational studies suggest that TNIK functions in the regulation of the cytoskeleton.
Specifications
TNIK | |
Polyclonal | |
Unconjugated | |
TNIK | |
1500031A17Rik; 4831440I19Rik; AI451411; C530008O15Rik; C630040K21Rik; KIAA0551; RGD1561817; Tnik; TRAF2 and NCK interacting kinase; TRAF2 and NCK-interacting protein kinase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100339162, 23043 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9UKE5 | |
TNIK | |
Synthetic 18 amino acid peptide from internal region of human TNIK. | |
50 μg | |
Primary | |
Human, Rabbit, Monkey, Bat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.