Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNIK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533086
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533086 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533086 Supplier Thermo Scientific Supplier No. PA533086
Add to Cart
Add to Cart

TNIK Polyclonal Antibody

TNIK Polyclonal Antibody for IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNIK
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene TNIK
Gene Accession No. Q9UKE5
Gene Alias 1500031A17Rik; 4831440I19Rik; AI451411; C530008O15Rik; C630040K21Rik; KIAA0551; RGD1561817; Tnik; TRAF2 and NCK interacting kinase; TRAF2 and NCK-interacting protein kinase
Gene Symbols TNIK
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from internal region of human TNIK.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23043
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.