Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNIK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116058
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116058 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116058 Supplier Invitrogen™ Supplier No. PA5116058
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TNIK.

TNIK or TRAF2 and NCK interacting kinase is characterized by an N-terminal kinase domain and a C-terminal GCK domain that serves a regulatory function. TNIK is mainly expression in brain, heart, and spleen and it is a specific effector of RAP2 which regulate actin cytoskeleton. TNIK is autophosphorylated in a manner dependent upon lys54 in the ATP-binding pocket of its kinase domain and plays a main role in cytoskeleton regulation. TNIK has also been shown to phosphorylate gelsolin, the principal intracellular and extracellular actin-severing protein, in vitro. This and evidence from mutational studies suggest that TNIK functions in the regulation of the cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNIK
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TNIK
Gene Accession No. P83510, Q9UKE5
Gene Alias 1500031A17Rik; 4831440I19Rik; AI451411; C530008O15Rik; C630040K21Rik; KIAA0551; RGD1561817; Tnik; TRAF2 and NCK interacting kinase; TRAF2 and NCK-interacting protein kinase
Gene Symbols TNIK
Host Species Rabbit
Immunogen A synthesized peptide derived from human TNIK(Accession Q9UKE5), corresponding to amino acid residues R663-R713.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23043, 665113
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.