Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNIP1 Monoclonal Antibody (1A11E3)
GREENER_CHOICE

Catalog No. 376100
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-610-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-610-0 Supplier Invitrogen™ Supplier No. 376100
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

TNIP1 Monoclonal Antibody for Western Blot, ELISA, IP

Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNIP1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1A11E3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene TNIP1
Gene Accession No. Q15025
Gene Alias A20-binding inhibitor of NF-kappa B activation; A20-binding inhibitor of NF-kappa-B activation 1; ABIN; ABIN1; ABIN-1; AU018810; HIV-1 Nef-interacting protein; KIAA0113; Naf1; Nef; Nef-associated factor 1; Nef-associated factor 1 SNP; nip40-1; TNFAIP3 interacting protein 1; TNFAIP3-interacting protein 1; Tnip1; VAN; Virion-associated nuclear shuttling protein
Gene Symbols TNIP1
Host Species Mouse
Immunogen Synthetic peptide derived from the N-terminal region of the human, mouse and rat ABIN-1 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10318
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.