Learn More
Invitrogen™ TNIP3 Polyclonal Antibody

Description
A suggested positive control is human spleen tissue lysate. PA5-20798 can be used with blocking peptide PEP-0912.
Specifications
Specifications
| Antigen | TNIP3 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | TNIP3 |
| Gene Accession No. | Q96KP6 |
| Gene Alias | 9030611K07Rik; A20-binding inhibitor of NF-kappa-B activation 3; ABIN3; ABIN-3; ABIN-3 beta; LIND; Listeria induced; listeria-induced gene protein; TNFAIP3 interacting protein 3; TNFAIP3-interacting protein 3; TNFAIP3-interacting protein 3 beta; Tnip3; TNIP3 beta |
| Gene Symbols | TNIP3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.