Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNK1 Monoclonal Antibody (1B5G3;3F11H6)
GREENER_CHOICE

Catalog No. PIMA515413
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515413 Supplier Invitrogen™ Supplier No. MA515413
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15413 targets TNK1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15413 immunogen is purified recombinant fragment of TNK1 (aa451-560) expressed in E. Coli. .

TNK1 tyrosine kinase, non-receptor 1 belongs to the tyrosine protein kinase family and as such is an important regulator of intracellular signal transduction pathways mediating cellular proliferation, survival, and development. TNK1 is highly expressed in fetal tissues and at lower levels in a few adult tissues. TNK1 may function in signaling pathways utilized broadly during fetal development, and more selectively in adult tissues. TNK1 plays a negative regulatory role in the Ras-Raf1-MAPK pathway, and knockout mice have been shown to develop spontaneous tumors, suggesting a role as a tumor suppressor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNK1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B5G3;3F11H6
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene TNK1
Gene Accession No. Q13470
Gene Alias CD38 negative kinase 1; Kinase of embryonic stem cells; Kos1; MGC46193; non-receptor tyrosine kinase; non-receptor tyrosine-protein kinase TNK1; TNK1; Tnk1a; Tnk1b; tyrosine kinase non receptor 1; tyrosine kinase, non-receptor 1; tyrosine kinase, non-receptor, 1
Gene Symbols TNK1
Host Species Mouse
Immunogen Purified recombinant fragment of TNK1 (aa451-560) expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8711
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.