Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNN Monoclonal Antibody (BC-24)
GREENER_CHOICE

Catalog No. PIMA532994
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532994 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532994 Supplier Invitrogen™ Supplier No. MA532994
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with 1 mL of PBS buffer to yield a concentration of 100 μg/mL. Positive Control - IHC: Human Mammary Cancer Tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Extracellular matrix protein that seems to be a ligand for ITGA8:ITGB1, ITGAV:ITGB1 and ITGA4:ITGB1 (PubMed:17909022). Involved in neurite outgrowth and cell migration in hippocampal explants. During endochondral bone formation, inhibits proliferation and differentiation of proteoblasts mediated by canonical WNT signaling. In tumors, stimulates angiogenesis by elongation, migration and sprouting of endothelial cells (PubMed:19884327). Expressed in most mammary tumors, may facilitate tumorigenesis by supporting the migratory behavior of breast cancer cells (PubMed:17909022). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNN
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone BC-24
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites, 1.2% sodium acetate with 2mg BSA and 0.01mg sodium azide
Gene TNN
Gene Accession No. Q9UQP3
Gene Alias tenascin N; tenascin-N; Tenascin-W; tenasin W; TNN; TN-N; Tnw; TN-W
Gene Symbols TNN
Host Species Mouse
Immunogen Human tenascin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63923
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.