Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555411
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555411 Supplier Invitrogen™ Supplier No. PA555411
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEQQVTVSHT YKIDVPKSAL VQVDADPQPL SDDGASLLAL GEAREEQNII FRHNIRLQTP QKDCELAGSV QDLLARVKKL EEEMVEMKEQ CSA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.

Extracellular matrix protein that seems to be a ligand for ITGA8:ITGB1, ITGAV:ITGB1 and ITGA4:ITGB1 (PubMed:17909022). Involved in neurite outgrowth and cell migration in hippocampal explants. During endochondral bone formation, inhibits proliferation and differentiation of proteoblasts mediated by canonical WNT signaling. In tumors, stimulates angiogenesis by elongation, migration and sprouting of endothelial cells (PubMed:19884327). Expressed in most mammary tumors, may facilitate tumorigenesis by supporting the migratory behavior of breast cancer cells (PubMed:17909022). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNN
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TNN
Gene Accession No. Q9UQP3
Gene Alias tenascin N; tenascin-N; Tenascin-W; tenasin W; TNN; TN-N; Tnw; TN-W
Gene Symbols TNN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TNN. Recombinant protein control fragment (Product #RP-94145).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63923
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.