Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNNC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560269
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560269 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560269 Supplier Invitrogen™ Supplier No. PA560269
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NADGYIDPEE LAEIFRASGE HVTDEEIESL M Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 97%.

Troponin (Tn), a key protein complex in the regulation of striated muscle contraction, is composed of 3 subunits. The Tn-I subunit inhibits actomyosin ATPase, the Tn-T subunit binds tropomyosin and Tn-C, while the Tn-C subunit binds calcium and overcomes the inhibitory action of the troponin complex on actin filaments. The protein encoded by this gene is the Tn-C subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNNC2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TNNC2
Gene Accession No. P02585
Gene Alias skeletal troponin C; STNC; Tncs; Tnnc2; troponin C type 2 (fast); troponin C, fast skeletal; troponin C, skeletal muscle; troponin C2, fast; troponin C2, fast skeletal type
Gene Symbols TNNC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TNNC2. Recombinant protein control fragment (Product #RP-99339).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7125
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.