Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TNNT3 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537899
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.
Specifications
TNNT3 | |
Polyclonal | |
Unconjugated | |
TNNT3 | |
beta TnTF; beta-TnTF; Fast skeletal muscle troponin T; fast skeletal muscle troponin T3; fast skeletal troponin T beta-1 isoform; fetal troponin T 1; fetal troponin T 2; fetal troponin T 3; fTnT; skeletal muscle fast-twitch TnT; skeletal troponin T alpha-1 isoform; Tnnt3; Tnt; TnT1f; TnT2.5f; TnT2f; TnT2fa; TnT4f; tnTf; troponin T 2.5f precursor; troponin T 2f precursor; troponin T 2fa precursor; troponin T 3f; troponin T 4f precursor; troponin T fast skeletal muscle type; troponin T type 3 (skeletal, fast); troponin T, fast skeletal muscle; troponin T1f precursor; troponin T3, fast skeletal type; troponin T3, skeletal, fast; troponin-T3, skeletal, fast | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
21957, 24838, 7140 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P09739, P45378, Q9QZ47 | |
TNNT3 | |
Peptide with sequence C-TTLRSRIDQAQKHSK, from the C-terminus of TNNT3 (aa 240-254). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction