Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TNNT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543681
Description
Peptide sequence: PRPKLTAPKI PEGEKVDFDD IQKKRQNKDL MELQALIDSH FEARKKEEEE Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.
Specifications
TNNT3 | |
Polyclonal | |
Unconjugated | |
TNNT3 | |
beta TnTF; beta-TnTF; Fast skeletal muscle troponin T; fast skeletal muscle troponin T3; fast skeletal troponin T beta-1 isoform; fetal troponin T 1; fetal troponin T 2; fetal troponin T 3; fTnT; skeletal muscle fast-twitch TnT; skeletal troponin T alpha-1 isoform; Tnnt3; Tnt; TnT1f; TnT2.5f; TnT2f; TnT2fa; TnT4f; tnTf; troponin T 2.5f precursor; troponin T 2f precursor; troponin T 2fa precursor; troponin T 3f; troponin T 4f precursor; troponin T fast skeletal muscle type; troponin T type 3 (skeletal, fast); troponin T, fast skeletal muscle; troponin T1f precursor; troponin T3, fast skeletal type; troponin T3, skeletal, fast; troponin-T3, skeletal, fast | |
Rabbit | |
Protein A | |
RUO | |
7140 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P45378 | |
TNNT3 | |
Synthetic peptide directed towards the N-terminal of human TNNT3 (aa 50-99). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction