Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNNT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114785
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114785 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114785 Supplier Invitrogen™ Supplier No. PA5114785
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TNNT3.

Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNNT3
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TNNT3
Gene Accession No. P45378, Q9QZ47
Gene Alias beta TnTF; beta-TnTF; Fast skeletal muscle troponin T; fast skeletal muscle troponin T3; fast skeletal troponin T beta-1 isoform; fetal troponin T 1; fetal troponin T 2; fetal troponin T 3; fTnT; skeletal muscle fast-twitch TnT; skeletal troponin T alpha-1 isoform; Tnnt3; Tnt; TnT1f; TnT2.5f; TnT2f; TnT2fa; TnT4f; tnTf; troponin T 2.5f precursor; troponin T 2f precursor; troponin T 2fa precursor; troponin T 3f; troponin T 4f precursor; troponin T fast skeletal muscle type; troponin T type 3 (skeletal, fast); troponin T, fast skeletal muscle; troponin T1f precursor; troponin T3, fast skeletal type; troponin T3, skeletal, fast; troponin-T3, skeletal, fast
Gene Symbols TNNT3
Host Species Rabbit
Immunogen A synthesized peptide derived from human TNNT3(Accession P45378), corresponding to amino acid residues Q220-K269.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21957, 7140
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.