Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TNP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544078
Description
Peptide sequence: MSTSRKLKSH GMRRSKSRSP HKGVKRGGSK RKYRKGNLKS RKRGDDANRN Sequence homology: Cow: 92%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 92%.
TNP1 is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines. Transition protein-1 is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines (see PRM1, MIM 182880; PRM2, MIM 182890) (Luerssen et al., 1990 ). Sequence Note: removed 4 bases from the 5' end that did not align to the reference genome assembly.
Specifications
| TNP1 | |
| Polyclonal | |
| Unconjugated | |
| Tnp1 | |
| Protein T; Spermatid nuclear transition protein 1; STP-1; Testis-specific basic protein; tnp; TNP1; TNP1 protein; TP1; TP-1; transition protein 1; transition protein 1 (during histone to protamine replacement) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7141 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P09430 | |
| Tnp1 | |
| Synthetic peptide directed towards the N-terminal of human TNP1 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction