Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560696
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560696 Supplier Invitrogen™ Supplier No. PA560696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STSRKLKSHG MRRSKSRSPH KGVKRGGSKR KYRKGNLKSR KRGDDANRNY RSHL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

TNP1 is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines. Transition protein-1 is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines (see PRM1, MIM 182880; PRM2, MIM 182890) (Luerssen et al., 1990 ). Sequence Note: removed 4 bases from the 5' end that did not align to the reference genome assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tnp1
Gene Accession No. P09430
Gene Alias Protein T; Spermatid nuclear transition protein 1; STP-1; Testis-specific basic protein; tnp; TNP1; TNP1 protein; TP1; TP-1; transition protein 1; transition protein 1 (during histone to protamine replacement)
Gene Symbols Tnp1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TNP1. Recombinant protein control fragment (Product #RP-92289).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7141
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.