Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103981
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103981 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103981 Supplier Invitrogen™ Supplier No. PA5103981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TNP2.

During mammalian spermiogenesis, histones are transiently replaced by several low molecular weight proteins called transition proteins (TNPs) . Transition proteins facilitate chromatin transformation from the nucleosome structure to the nucleoprotamine structure during spermatid differentiation . Transition protein-2, also known as TNP2, and TP2, maps to human chromosome 16p13.13 and encodes a highly basic nuclear protein. TNP2 is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines . TNP2 is not a critical factor for shaping of the sperm nucleus, histone displacement, initiation of chromatin condensation, binding of protamines to DNA, or fertility . However, TNP2 is necessary for maintaining the normal processing of protamine 2 and, consequently, the completion of chromatin condensation. If TNP1 is missing, TNP2 may partially compensate for TNP1, but this dysregulation of nucleoprotein replacement results in an abnormal pattern of chromatin condensation and in reduced fertility .
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TNP2
Gene Accession No. Q05952
Gene Alias haploid expressed, male germcell specific, chromatin binding; nuclear transition protein 2; TNP2; TP2; TP-2; transition protein 2; transition protein 2 (during histone to protamine replacement)
Gene Symbols TNP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TNP2(Accession Q05952), corresponding to amino acid residues Q39-H89.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7142
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.