Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30444220UL
Description
TNP2 Polyclonal specifically detects TNP2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
TNP2 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
7142 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
nuclear transition protein 2, TP2, transition protein 2 (during histone to protamine replacement) | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-138 of human TNP2 (NP_005416.1). | |
20 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction