Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNPO1 Monoclonal Antibody (D45)
GREENER_CHOICE

Catalog No. PIMA546555
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546555 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546555 Supplier Invitrogen™ Supplier No. MA546555
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TNPO1 Monoclonal Antibody for Western Blot, ICC/IF, IP, ELISA

TNPO1 comprises the beta subunit of the karyopherin receptor complex which interacts with nuclear localization signals to target nuclear proteins to the nucleus. The karyopherin receptor complex is a heterodimer of an alpha subunit which recognizes the nuclear localization signal and a beta subunit which docks the complex at nucleoporins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNPO1
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone D45
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TNPO1
Gene Accession No. Q8BFY9, Q92973
Gene Alias AU021749; D13Ertd688e; importin 2; importin beta 2; importin beta-2; IPO2; karyopherin (importin) beta 2; karyopherin beta-2; KPNB2; M9 region interaction protein; MAPKAP1; MIP; MIP1; MSIN1; SAPK 1; SIN1; SIN-1; SIN1g; TNPO1; transportin; transportin 1; transportin-1; TRN; Unknown (protein for MGC:127455)
Gene Symbols TNPO1
Host Species Mouse
Immunogen Recombinant Human his tagged transportin1.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100343169, 238799, 3842
Target Species Human, Mouse, Rabbit, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.