Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNPO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116066
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116066 Supplier Thermo Scientific Supplier No. PA5116066
Add to Cart
Add to Cart

TNPO1 Polyclonal Antibody

TNPO1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNPO1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene TNPO1
Gene Accession No. Q8BFY9, Q92973
Gene Alias AU021749; D13Ertd688e; importin 2; importin beta 2; importin beta-2; IPO2; karyopherin (importin) beta 2; karyopherin beta-2; KPNB2; M9 region interaction protein; MAPKAP1; MIP; MIP1; MSIN1; SAPK 1; SIN1; SIN-1; SIN1g; TNPO1; transportin; transportin 1; transportin-1; TRN; Unknown (protein for MGC:127455)
Gene Symbols TNPO1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TNPO1(Accession Q92973), corresponding to amino acid residues P853-V898.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238799, 309126, 3842
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.