Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNPO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567176 Supplier Invitrogen™ Supplier No. PA567176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDIKPRFHKSR TVTLPHEAER PDGSEDAEDD DD Highest antigen sequence identity to the following orthologs - mouse 94%, rat 97%.

Probably functions in nuclear protein import as nuclear transport receptor. Serves as receptor for nuclear localization signals (NLS) in cargo substrates. Is thought to mediate docking of the importin/substrate complex to the nuclear pore complex (NPC) through binding to nucleoporin and the complex is subsequently translocated through the pore by an energy requiring, Ran-dependent mechanism. At the nucleoplasmic side of the NPC, Ran binds to the importin, the importin/substrate complex dissociates and importin is re-exported from the nucleus to the cytoplasm where GTP hydrolysis releases Ran. The directionality of nuclear import is thought to be conferred by an asymmetric distribution of the GTP- and GDP-bound forms of Ran between the cytoplasm and nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNPO2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TNPO2
Gene Accession No. O14787
Gene Alias 1110034O24Rik; AA414969; AI464345; AI852433; importin 3; IPO3; karyopherin (importin) beta 2b; karyopherin beta 2b, transportin; karyopherin beta-2b; Knpb2b; Kpnb2b; TNPO2; transportin 2; transportin 2 (importin 3, karyopherin beta 2b); transportin-2; TRN2
Gene Symbols TNPO2
Host Species Rabbit
Immunogen Recombinant Human TNPO2. Recombinant protein control fragment (Product #RP-107820).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30000
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.