Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNPO3 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176374MFV610
Description
TNPO3 Polyclonal antibody specifically detects TNPO3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
TNPO3 | |
Polyclonal | |
Western Blot, ELISA | |
imp12, MTR10A, transportin 3, transportin-3 | |
Antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human TNPO3. The immunogen is located within amino acids 220 - 270 of TNPO3. Amino Acid Squence: EVLQQDKSTSSNLHEAASD | |
0.1 mL | |
Signal Transduction | |
23534 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction