Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNPO3 Recombinant Rabbit Monoclonal Antibody (0U3Q1)
GREENER_CHOICE

Catalog No. PIMA537991
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537991 Supplier Invitrogen™ Supplier No. MA537991
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, RD, Jurkat, Mouse lung, Mouse testis, Mouse thymus, Rat testis Immunogen sequence: QVMNQLGQQL VSQLLHTCCF CLPPYTLPDV AEVLWEIMQV DRPTFCRWLE NSLKGLPKET TVGAVTVTHK QLTDFHKQVT SAEECKQVCW ALRDFTRLFR.

TNPO3 is a member of the importin beta/transportin family of nuclear import receptors and is specific for the essential splicing factors termed SR proteins, binding specifically and directly to the arginine/serine (RS)-rich domains of these proteins. Further experiments demonstrated that this interaction occurs only when these RS domains are phosphorylated. More recently, TNPO3 was identified as an HIV dependency factor (HDF) and that depletion of TNPO3 resulted in HIV inhibition after reverse transcription but before integration, suggesting that TNPO3 may be an important drug target in HIV treatment. At least three isoforms of TNPO3 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNPO3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 0U3Q1
Concentration 1.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TNPO3
Gene Accession No. Q6P2B1, Q9Y5L0
Gene Alias 5730544L10Rik; C430013M08Rik; C81142; D6Ertd313e; Imp12; importin 12; Importin-12; IPO12; LGMD1F; limb girdle muscular dystrophy 1F (autosomal dominant); mKIAA4133; MTR10A; Tnpo3; transportin 3; transportin-3; Transportin-SR; TRNSR; TRN-SR; TRN-SR2
Gene Symbols TNPO3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 824-923 of human Transportin 3 (Transportin 3 (TNPO3)) (Q9Y5L0).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23534, 296954, 320938
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.