Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TNRC4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552272
Description
Immunogen sequence: PVSAIPAALG VNGYSPVPTQ PTGQPAPDAL YPNGVHPYPA QSPAAPVDPL QQAYAGMQHY TAAYPAAYSL VAPAFPQPPA LVAQQPPPPP QQQQQQQQQQ QQQQQREGPD GCNIFIYHLP QEFTDSEILQ MFV Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 94%.
RNA-binding protein involved in the regulation of pre- mRNA alternative splicing. Mediates exon inclusion and/or exclusion in pre-mRNA that are subject to tissue-specific and developmentally regulated alternative splicing. Specifically activates exon 5 inclusion of cardiac isoforms of TNNT2 during heart remodeling at the juvenile to adult transition. Activates the splicing of MAPT/Tau exon 10. Binds to muscle-specific splicing enhancer (MSE) intronic sites flanking the alternative exon 5 of TNNT2 pre-mRNA.
Specifications
TNRC4 | |
Polyclonal | |
Unconjugated | |
CELF3 | |
4930415M08Rik; BRUNOL1; Bruno-like protein 1; CAG repeat domain; CAG repeat protein 4; CAGH4; CELF3; CELF-3; CUG-BP and ETR-3 like factor 3; CUG-BP- and ETR-3-like factor 3; CUGBP Elav-like family member 3; CUGBP, Elav-like family member 3; ELAV-type RNA-binding protein 1; ERDA4; ETR-1; Expanded repeat domain protein CAG/CTG 4; expanded repeat domain, CAG/CTG 4; RGD1563168; RNA-binding protein BRUNOL-1; Tnrc4; trinucleotide repeat containing 4; trinucleotide repeat-containing gene 4 protein; VCUGBP, Elav-like family member 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11189 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5SZQ8 | |
CELF3 | |
Recombinant protein corresponding to Human TNRC4. Recombinant protein control fragment (Product #RP-89180). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction