Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNRC6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553650 Supplier Invitrogen™ Supplier No. PA553650
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLQRLLAQQQ RAQSQRSVPS GNRPQQDQQG RPLSVQQQMM QQSRQLDPNL LVKQQTPPSQ QQPLHQPAMK SFLDNVMPHT TPELQKGPSP INAFSNFPIG LNSNLNVNMD MNSIKEPQSR LRKWT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

TNRC6A is a member of the trinucleotide repeat containing 6 protein family. The protein functions in post-transcriptional gene silencing through the RNA interference (RNAi) and microRNA pathways. The protein associates with messenger RNAs and Argonaute proteins in cytoplasmic bodies known as GW-bodies or P-bodies. Inhibiting expression of this gene delocalizes other GW-body proteins and impairs RNAi and microRNA-induced gene silencing.This gene encodes a member of the trinucleotide repeat containing 6 protein family. The protein functions in post-transcriptional gene silencing through the RNA interference (RNAi) and microRNA pathways. The protein associates with messenger RNAs and Argonaute proteins in cytoplasmic bodies known as GW-bodies or P-bodies. Inhibiting expression of this gene delocalizes other GW-body proteins and impairs RNAi and microRNA-induced gene silencing. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNRC6A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TNRC6A
Gene Accession No. Q8NDV7
Gene Alias 2010321I05Rik; 3110054G10Rik; AW557223; CAG repeat protein 26; CAGH26; CTD-2540M10.1; D130023A07Rik; DKFZp666E117; EDIE; EMSY interactor protein; FLJ22043; Glycine-tryptophan protein of 182 kDa; GW1; GW182; GW182 autoantigen; KIAA1460; MGC75384; Protein GW1; Tnrc6; TNRC6A; trinucleotide repeat containing 6; trinucleotide repeat containing 6A; trinucleotide repeat containing adaptor 6A; trinucleotide repeat-containing gene 6A protein
Gene Symbols TNRC6A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TNRC6A. Recombinant protein control fragment (Product #RP-91607).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27327
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.