Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNRC6A Polyclonal Antibody
TNRC6A Polyclonal Antibody
Supplier: Thermo Scientific PA576194
Description
TNRC6A Polyclonal Antibody for Western Blot, ICC/IF

Specifications
TNRC6A | |
Polyclonal | |
Unconjugated | |
TNRC6A | |
2010321I05Rik; 3110054G10Rik; AW557223; CAG repeat protein 26; CAGH26; CTD-2540M10.1; D130023A07Rik; DKFZp666E117; EDIE; EMSY interactor protein; FLJ22043; Glycine-tryptophan protein of 182 kDa; GW1; GW182; GW182 autoantigen; KIAA1460; MGC75384; Protein GW1; Tnrc6; TNRC6A; trinucleotide repeat containing 6; trinucleotide repeat containing 6A; trinucleotide repeat containing adaptor 6A; trinucleotide repeat-containing gene 6A protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
233833, 27327, 308971 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3UHK8, Q8NDV7 | |
TNRC6A | |
Synthetic peptide of human TNRC6A. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction