Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNRC6A Polyclonal Antibody, Invitrogen™

Catalog No. PIPA541895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541895 Supplier Thermo Scientific Supplier No. PA541895

Rabbit Polyclonal Antibody

Peptide sequence: RELEAKATKD VERNLSRDLV QEEEQLMEEK KKKKDDKKKK EAAQKKATEQ Sequence homology: Dog: 93%; Horse: 100%; Human: 100%; Mouse: 100%

TNRC6A Polyclonal antibody specifically detects TNRC6A in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNRC6A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TNRC6A
Gene Accession No. Q8NDV7
Gene Alias 2010321I05Rik; 3110054G10Rik; AW557223; CAG repeat protein 26; CAGH26; CTD-2540M10.1; D130023A07Rik; DKFZp666E117; EDIE; EMSY interactor protein; FLJ22043; Glycine-tryptophan protein of 182 kDa; GW1; GW182; GW182 autoantigen; KIAA1460; MGC75384; Protein GW1; Tnrc6; TNRC6A; trinucleotide repeat containing 6; trinucleotide repeat containing 6A; trinucleotide repeat containing adaptor 6A; trinucleotide repeat-containing gene 6A protein
Gene Symbols TNRC6A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TNRC6A
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27327
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.