Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TNRC6B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541742
Description
Peptide sequence: LQSESGTAPV WSKSTPPAPD NGTSAWGEPN ESSPGWGEMD DTGASTTGWG Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The function of TNRC6B remains unknown.
Specifications
TNRC6B | |
Polyclonal | |
Unconjugated | |
TNRC6B | |
2700090M07Rik; A730065C02Rik; AI848765; androgen receptor-related apoptosis-associated protein CBL27; Cbl27; D230019K20Rik; KIAA1093; TNRC6B; trinucleotide repeat containing 6B; trinucleotide repeat containing adaptor 6B; trinucleotide repeat-containing gene 6B protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23112 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UPQ9 | |
TNRC6B | |
Synthetic peptide directed towards the N-terminal of human TNRC6B. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction