Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNRC6B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981408
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1408 Supplier Bioss Supplier No. BS11489R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function of TNRC6B remains unknown.

Specifications

Antigen TNRC6B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TNRC6B
Gene Accession No. Q9UPQ9
Gene Alias 2700090M07Rik; A730065C02Rik; adenylosuccinate lyase; AI848765; androgen receptor-related apoptosis-associated protein CBL27; BOS_6138; Cbl27; D230019K20Rik; Kiaa1093; LOC100337846; Tnrc6b; trinucleotide repeat containing 6B; Trinucleotide repeat-containing gene 6B protein
Gene Symbols TNRC6B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TNRC6B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23112
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.