Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TNXB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580173
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580173 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580173 Supplier Invitrogen™ Supplier No. PA580173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: MCF-7 whole cell, SW620 whole cell, HELA whole cell. IHC: human placenta tissue. ICC/IF: MCF7 cell. Flow: Hela cell.

Tenascin-X (TNXB) is a extracellular matrix glycoprotein predominantly located in the outer reticular lamina of the basement membrane (BM). It interacts with many other ECM proteins and it accelerates collagen fibrillogenesisin vitro. TNXB plays a role in interactions between cell and ECM, antiadhesive effect, inhibiting cell migration, and in maintaining homeostasis of the extracellular matrix. Deficiency of TNXB has been associated with the connective tissue disorder Ehlers-Danlos syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNXB
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene TNXB
Gene Accession No. P22105
Gene Alias EDS3; growth-inhibiting protein 45; hexabrachion-like protein; HXBL; tenascin XB; tenascin XB1; tenascin XB2; tenascin-X; TENX; Tn-mhc; Tnx; TN-X; TNXB; TNXB1; TNXB2; TNXB2tenascin XB1; TNXBS; TNXBSTNXTENX; VUR8; XB; XBS; XB-S; XBTN-X
Gene Symbols TNXB
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human TNXB (3309-3337aa DLRAVAVSGLDPARKYKFLLFGLQNGKRH).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7148
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.