Learn More
Invitrogen™ TO-PRO™3 Ready Flow™ Reagent
Description
- A bright and easy-to-use nucleic acid stain for distinguishing dead cells from live cells in flow cytometry assays
- Dye has excitation/emission maxima of 642/661 nm, similar to Alexa Fluor™ 647 or allophycocyanin (APC)
- This direct-to-sample dead-cell stain emits red fluorescence when bound to DNA
- Add 2 drops form the dropper bottle directly into 1 x 106 cells, incubate, and analyze
- No need to dilute, weigh, or pipette
- Stable at room temperature
Specifications
Specifications
| Cell Permeability | Impermeant |
| Color | Far-red |
| Content And Storage | Includes 2 dropper bottles, store at room temperature. |
| Description | TO-PRO™3 Ready Flow™ Reagent |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Reagent |
| Dye Type | Cell-Impermeant |
| Form | Solution |
| Show More |
Frequently Asked Questions (FAQs)
Yes, you may scale up or down as needed, but we recommend keeping the cell density the same. There are 41 µL per drop, so 2 drops is 82 µL (for the 1 mL of sample). You need to only scale down for this 100 µL volume, which would be 8.2 µL.
The amount of dye or reagent in the Ready Flow products is proprietary. If you need to consider the exact amount of dye or reagent for your experiment, each of the Ready Flow reagents is available as a standalone product.
This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.