Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565480
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565480 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565480 Supplier Invitrogen™ Supplier No. PA565480
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STKMKNSGRSN KVARTSPINL GLNVNDLLKQ KAISSSMHSL YGLGLGSQQQ PQQQ Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.

This gene encodes a member of the tob/btg1 family of anti-proliferative proteins that have the potential to regulate cell growth. When exogenously expressed, this protein supresses cell growth in tissue culture. The protein undergoes phophorylation by a serine/threonine kinase, 90 kDa ribosomal S6 kinase. Interactions of this protein with the v-erb-b2 erythroblastic leukemia viral oncogene homolog 2 gene product p185 interferes with growth suppression. This protein inhibits T cell proliferation and transcription of cytokines and cyclins. The protein interacts with both mothers against decapentaplegic Drosophila homolog 2 and 4 to enhance their DNA binding activity. This interaction inhibits interleukin 2 transcription in T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOB1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TOB1
Gene Accession No. P50616
Gene Alias APRO5; APRO6; MGC104792; MGC34446; PIG49; proliferation-inducing gene 49; protein Tob1; Protein Tob1-like protein; TOB; TOB1; transducer of erbB2; Transducer of erbB-2 1; transducer of ERBB2, 1; transducer of ErbB-2.1; Trob; TROB1; Unknown (protein for MGC:134447)
Gene Symbols TOB1
Host Species Rabbit
Immunogen Recombinant Human TOB1. Recombinant protein control fragment (Product #RP-106155).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10140
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.