Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562923
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562923 Supplier Invitrogen™ Supplier No. PA562923
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVASSGAGGQ QPPQQPRMAR SPTNSLLKHK SLS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.

TOB2 is a 344 amino acid rich novel member of the Tob/BTG1 anti-proliferative family of proteins characterized by similarities in their N-terminal BTG/Tob homology domains. This anti-proliferative protein inhibits cell cycle progression from the G0/G1 to S phases and is involved in cell cycle regulation through interaction with Caf1, a component of the CCR4 transcription factor complex that associates with cyclin-dependent kinases. Recent reports suggest that TOB2 plays important roles in spermatogenesis, embryonic dorsoventral patterning, osteogenesis, T-cell activation, learning and memory and acts primarily as a transcriptional repressor in several signaling pathways. TOB2, a signaling protein, may participate in transcriptional regulation of several genes. It is a regulatory protein ubiquitously expressed in most of the tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOB2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tob2
Gene Accession No. Q14106
Gene Alias 2900090N22Rik; 4930545K18Rik; APRO5; AV071822; hypothetical protein; KIAA1663; mKIAA1663; protein Tob2; Protein Tob4; Tob2; TOB4; TOBL; Transducer of erbB-2 2; transducer of ERBB2, 2; TROB2
Gene Symbols Tob2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TOB2. Recombinant protein control fragment (Product #RP-101553).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10766
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.