Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tobramycin, 900μg/mg

Catalog No. p-7076640
Click to view available options
Quantity:
1 g
5 g

CAS: 32986-56-4 | C18H37N5O9 | 467.52 g/mol

Primarily targets Gram-negative bacteria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32986-56-4
Molecular Formula C18H37N5O9
Molecular Weight (g/mol) 467.52
MDL Number MFCD00077885
InChI Key NLVFBUXFDBBNBW-PBSUHMDJSA-N
Synonym tobramycin, nebramycin 6, nebramycin factor 6, nebramycin vi, tobramicin, tobramycetin, nebcin, 3'-deoxykanamycin b, aktob, tobi
PubChem CID 36294
ChEBI CHEBI:28864
IUPAC Name (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N

Specifications

Packaging Plastic bottle
Quantity 1 g
Beilstein 1357507
Solubility Information Soluble in water
Formula Weight 467.51
Physical Form Powder
Chemical Name or Material Tobramycin
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 251-322-5
RTECSNumber WK2100000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.