Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOC Standard, According to ISO/CEN EN1484, 1,000 mg/L±10 mg/L, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
250 mL
500 mL
Specifications
Specifications
Concentration | 1000 mg/L ±10 mg/L |
Quantity | 250 mL |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction