Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOCA-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520445
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520445 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520445 Supplier Invitrogen™ Supplier No. PA520445
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-20445 can be used with blocking peptide PEP-0562.

The protein encoded by this gene contains a RING finger motif. The mouse counterpart of this protein has been shown to interact with Rela, the p65 subunit of NF-kappaB (NFKB), and modulate NFKB-mediated transcription activity. The mouse protein also binds ubiquitin-conjugating enzymes (E2s) and is a substrate for E2-dependent ubiquitination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOCA-1
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene FNBP1L
Gene Accession No. Q2HWF0, Q5T0N5, Q8K012
Gene Alias 2610318I01Rik; AW548221; C1orf39; Cdc42 effector; FNBP1L; formin binding protein 1 like; formin binding protein 1-like; formin-binding protein 1-like; RGD1305386; TOCA1; Toca-1; transducer of Cdc42-dependent actin assembly 1; transducer of Cdc42-dependent actin assembly protein 1
Gene Symbols FNBP1L
Host Species Rabbit
Immunogen A 15 amino acid peptide near the amino terminus of the human TOCA-1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 214459, 310839, 54874
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.