Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toca-1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501565221
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5221 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5221 Supplier Bethyl Laboratories, Inc Supplier No. A303470AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Actin reorganization is important for the regulation of neuronal morphology. A protein involved in this process, the transducer of cdc42-dependent actin assembly 1 (TOCA-1) protein, a member of the evolutionarily conserved pombe CDC15 homology (PCH) protein family, is an essential component of the Cdc42 pathway. TOCA-1 binds both N-WASP and Cdc42 and is essential for Cdc42- and PIP2-induced actin polymerization, suggesting that TOCA-1 mediates Cdc42-dependent actin nucleation by activating the N-WASP-WIP complex. Decreased expression of TOCA-1 significantly enhances neurite elongation in PC-12 cells; its overexpression in the same cells suppresses neurite elongation. It has been suggested that TOCA-1 negatively regulates axon branching by regulating membrane trafficking by regulating membrane trafficking through the F-BAR/EFC domain. Multiple isoforms of TOCA-1 are known to exist.

Specifications

Antigen Toca-1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene FNBP1L
Gene Accession No. Q5T0N5
Gene Alias C1orf39, TOCA1; formin-binding protein 1-like; toca-1; transducer of Cdc42-dependent actin assembly 1; transducer of Cdc42-dependent actin assembly protein 1
Gene Symbols FNBP1L
Host Species Rabbit
Immunogen Between 450 and 500
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54874
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.