Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tocilizumab Chimeric Recombinant Rabbit Monoclonal Antibody (rhPM-1 (Tocilizumab))
GREENER_CHOICE

Catalog No. PIMA548062
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548062 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548062 Supplier Invitrogen™ Supplier No. MA548062
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: Tocilizumab binds specifically to both the membrane bound and soluble forms of human IL-6R. The IL6 receptor is a protein complex consisting of an IL-6 receptor subunit (IL6R) and interleukin 6 signal transducer Glycoprotein 130. Interleukin 6 (IL6) is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in immune response.

Tocilizumab (Actemra) is a humanized monoclonal antibody that is developed by Hoffmann-La Roche and Chugai for the treatment of patients with rheumatoid arthritis (RA) and systemic juvenile idiopathic arthritis, a severe form of arthritis in children. Tocilizumab binds to the interleukin-6 receptor (IL-6R) and blocks its interaction with interleukin 6 (IL-6), hindering IL-6 from exerting its pro-inflammatory effects.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tocilizumab Chimeric
Applications ELISA, Inhibition Assays, Immunocytochemistry
Classification Recombinant Monoclonal
Clone rhPM-1 (Tocilizumab)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias Actemra; Atlizumab; MRA; R-1569; RHPM-1; RO-4877533
Host Species Rabbit
Immunogen This antibody was prepared by CDR grafting of mouse PM-1 antibody onto a human framework regions.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.