Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Amyloid beta-peptide (1-42) (human)

Click to view available options
Quantity:
100 μg
Description
Amyloid β-Peptide (1-42) (human) is a human form of the predominant amyloid β-peptide found in the brains of patients with Alzheimer's disease. Downregulates bcl-2 and increases the levels of bax. Neurotoxic.
Control Peptide also available.
Specifications
Specifications
Purity | 0.95 |
Content And Storage | Store at -20°C |
Quantity | 100 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction