Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Amyloid beta-peptide (1-42) (rat)
Click to view available options
Quantity:
500 μg
Description
Amyloid β-peptide (1-42) (rat) is a rat form of the predominant amyloid β-peptide found in plaques associated with Alzheimer's disease. Leads to a large shift in the bax/bcl-2 ratio in favour of pro-apoptotic bax. Neurotoxic.
Specifications
Specifications
Purity | 95% |
Content And Storage | Store at -20°C |
Quantity | 500 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction