Learn More
Tocris Bioscience™ DADLE
Click to view available options
Quantity:
5 mg
Description
DADLE is a prototypical δ-opioid receptor agonist that also displays activity at the μ-opioid receptor. Displays antinociceptive activity in vivo.
Specifications
Specifications
Purity | 0.95 |
Quantity | 5 mg |