Learn More
Tocris Bioscience™ Dynamin inhibitory peptide
Click to view available options
Quantity:
1 mg
Description
Dynamin inhibitory peptide is a peptide inhibitor of the GTPase dynamin; competitively blocks binding of dynamin to amphiphysin thus prevents endocytosis when administered intracellularly. Reduces GABAA receptor internalization and increases miniature ISPC amplitude and frequency in neurons expressing GABAA receptors.
Cell-permeable version also available.
Specifications
Specifications
Common Name | Dynamin Inhibitors |
Quantity | 1 mg |